Express Eingabe
Austria
AuftragsverfolgungKontakt
Natriumcaprinat ≥99%

Natriumcaprinat ≥99%

Glass, amber, 25 g
Catalog # SPCMD2852-25GM
Supplier:   Spectrum Chemical
Natriumcaprinat ≥99%
Catalog # SPCMD2852-25GM
Supplier:  Spectrum Chemical
CAS Number:  1002-62-6
Natriumcaprinat ≥99%

Glass, amber, 25 g

CAS Number:  1002-62-6
Price
66,50 €
Log in for availability
1 * 25 g
Product Details & Documents

Caution: Storage temperature - Refrigerate

  • CAS Number:1002-62-6
  • Formula:C₁₀H₁₉NaO₂
  • MDL Number:MFCD00066453
  • Molecular Weight:194.2464 g/mol
  • Storage Temperature:Kühlschrank
Specifications

Specification Test Results

  • Assay:
    Approx. 99 %

Specifications

  • Packung:
    Glass, amber
  • VE:
    25 g

Specifications