Connectors, click-on in-line super clean®
Catalog # REST22475
Supplier: Restek
Specifications
- Bezeichnung:Click-on in-line super clean connectors, Includes 3,175 mm brass
- Seq:01
- Environmentally Preferable:
- VE:2
Specifications
About this item
Click-on connectors allow you to change traps quickly, without introducing oxygen into your system.
- Each connector is 60 mm in length