C.L.E.D. agar with bromothymol blue dehydrated culture media
Catalog # HIMEM792-500G
Supplier: HIMEDIA
CAS Number:
Specifications
- VE:500 g
- Bezeichnung:C.L.E.D. agar with bromothymol blue
- Seq:01
Specifications
About this item
For the isolation and differentiation of urinary pathogens on the basis of lactose fermentation.
- Microbiological culture media
- 500 g poly bottles