Express Eingabe
Austria
AuftragsverfolgungKontakt
Magnesium Sulfate, Reference Standard | Avantor

Magnesium Sulfate, Reference Standard

Magnesium Sulfate, USP reference standard, Glasflasche für Feststoffe, 1 g
Artikelnr. 1374361.
Anbieter:   USP
Magnesium Sulfate, Reference Standard
Artikelnr. 1374361.
Anbieter:  USP
CAS Nummer:  
Magnesium Sulfate, Reference Standard
Magnesium Sulfate, Reference Standard

Magnesium Sulfate, USP reference standard, Glasflasche für Feststoffe, 1 g

CAS Nummer:  
Preis
387,00 €
Einloggen für Verfügbarkeit
1 * 1 g
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Zertifizierung: US Pharmacopea Reference Standard

Spezifikationen

Spezifikationen

  • Packung:
    Glasflasche für Feststoffe
  • VE:
    1 g
  • Normen:
    USP reference standard
  • Bezeichnung:
    Magnesium Sulfate
  • Cat. No.:
    1374361.
  • CAS Nr.:
    10034-99-8
  • Synonym(s):
    Magnesium Sulfate (1:1) heptahydrate
  • Bezeichnung:
    Magnesium Sulfate

Spezifikationen

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.