Lufenuron, Reference Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
- Catalog No:
- 1370724.
- 1370735.
- 1370779.
- 1370780.
- Bezeichnung:
- Lufenuron Related Compound B
- Lufenuron Related Compound C
- Lufenuron
- Lufenuron Related Compound G
- Synonym(s):
- N-[(2,5-Dichloro-4-hydroxyphenyl)carbamoyl]-2,6-difluorobenzamide
- N-[3-Chloro-4-(1,1,2,3,3,3-hexafluoropropoxy)phenylcarbamoyl]-2,6-difluorobenzamide
- 2,5-Dichloro-4-[3-(2,6-difluorobenzoyl)ureido]phenyl phenyl carbonate
- CAS Nr.:
- 103055-07-8
- UN No.:
- 3077
- 3077
Product Family Options
- BezeichnungNormenPackungVE
- Lufenuron Related Compound BUSP reference standardVial15 mg1 * 15 mg1 270,00 €
Specifications
Cat. No.1370724.BezeichnungLufenuron Related Compound BSynonym(s)N-[(2,5-Dichloro-4-hydroxyphenyl)carbamoyl]-2,6-difluorobenzamideCAS Nr.UN No. - Lufenuron Related Compound CUSP reference standardVial15 mg1 * 15 mg1 270,00 €
Specifications
Cat. No.1370735.BezeichnungLufenuron Related Compound CSynonym(s)N-[3-Chloro-4-(1,1,2,3,3,3-hexafluoropropoxy)phenylcarbamoyl]-2,6-difluorobenzamideCAS Nr.UN No. - LufenuronUSP reference standardVial200 mg1 * 200 mg378,00 €
Specifications
Cat. No.1370779.BezeichnungLufenuronSynonym(s)CAS Nr.103055-07-8UN No.3077 - Lufenuron Related Compound GUSP reference standardVial15 mg1 * 15 mg1 270,00 €
Specifications
Cat. No.1370780.BezeichnungLufenuron Related Compound GSynonym(s)2,5-Dichloro-4-[3-(2,6-difluorobenzoyl)ureido]phenyl phenyl carbonateCAS Nr.UN No.3077