Graphite O-ring for Shimadzu 2010 and 2014 GCs
Catalog # REST20243
Supplier: Restek
Specifications
- Bezeichnung:O-ring for Shimadzu 2010 and 2014 GCs
- Material:Graphit
- VE:5
Specifications
About this item
Graphite O-ring are for use with numerous Shimadzu gas chromatography systems.
- Available in packs of 5
- Durable graphite construction
- Max operating temperature of 450 °C
For use with Shimadzu 2010 and 2014 GC systems.