Fixed-Angle Titanium Rotor Package, Type 42.2 Ti, Optima
The Type 42.2 Ti rotor package, rated for 42000 rpm (min⁻¹), is a fixed angle rotor designed to centrifuge up to 72 tubes at a 30° angle to the axis of rotation. Used in Beckman Coulter class H, R, and S preparative ultracentrifuges, the rotor develops centrifugal forces that can efficiently separate intact viruses, microsomes, and lipoproteins in small-volume samples.
For use in instruments classified: HRS; Includes: (BECM342303) Cellulose propionate tubes (bag of 100 each), (BECM330329) Overspeed disk 42000 rpm (min⁻¹), (BECM332688) Vise assembly, (BECM343008) Spanner wrench, (BECM878446) Forceps, (BECM306812) Spinkote lubricant, (BECM335148) Vacuum grease.
Note: Originally this rotor was intended for small-sample lipoprotein applications and was called the LP 42 Ti. However, it has become a popular choice for processing a variety of microsamples, and the name has been changed to the Type 42.2 Ti to reflect its more general use.
- Bezeichnung:Type 42.2 Ti fixed angle titanium rotor package
- Für:Fixed angle (ultracentrifugation)
- Cat. No.:521-3750
- Radius:113 mm
- Max. RPM:42000