High Volume Chemical Resistant Siphon Pump, 5 strokes per gallon, 2" IPS adapter
Catalog # MFLX06433-72
Supplier: Avantor Fluid Handling
Specifications
- Förderleistung:5 strokes/gal.
- Bezeichnung:High Volume Chemical Resistant Siphon Pump, 5 strokes per gallon, 2" IPS adapter
- Environmentally Preferable:
- Wetted materials:Solvent-welded PVC body and shaft;vinyl hose; Viton valves and seals
- Tube length, inlet:38"
- Garantie:1 Month
- VE:1
Specifications
About this item
Ideal for most acids, bases, alcohols, aldehydes and aliphatic hydrocarbons.
- PVC with Viton® valves and seals for chemical resistance
- Withstands temperatures upto 120F (49C)
- Self-priming
This hand-operated drum pump is ideal for pumping most acids, bases, alcohols, aldehydes and aliphatic hydrocarbons. Constructed of PVC with Viton® valves and seals for optimal chemical resistance, this pump offers 5 strokes/gallon.
- Förderleistung:5 strokes/gal.
- Bezeichnung:High Volume Chemical Resistant Siphon Pump, 5 strokes per gallon, 2" IPS adapter
- Environmentally Preferable:
- Wetted materials:Solvent-welded PVC body and shaft;vinyl hose; Viton valves and seals
- Tube length, inlet:38"
- Garantie:1 Month
- VE:1