7000 Safety spectacle, Junior Stealth™
Artikelnr. JSPAASA918-321-100
Lieferant: JSP
Spezifikationen
- Bezeichnung:Junior Stealth™ 7000 Safety spectacle
- Verglasung:Clear PC
- VE:1
Spezifikationen
Über diesen Artikel
Contour design and flexibility of frame give a good versatile fit onto both smaller and larger faces.
- One-piece lens
- Permanently attached side arms for durability
- Optical class 1
The one-piece lens features permanently attached side arms for durability. Lens and frame offer protection against low energy, high-speed particles at 45 m/s (101 mph). The optical qualities of the lens comply with the requirements of optical class 1 (highest).
Certifications: Conforms to EN 166.