Trace elements C, Corning®
Catalog # MDTC25-023-CI
Supplier: Corning
CAS Number:
Specifications
- VE:100 mL
- Bezeichnung:Trace elements C
Specifications
About this item
Trace Elements C Solution has the same composition as Cleveland's Trace Elements I, and contains the trace elements found in the serum-free medium MCDB301.