Hink's TNM-FH medium, Corning®
Artikelnr. MDTC13-100-CV
Lieferant: Corning
CAS-Nummer:
Spezifikationen
- Medienformat:Flüssigkeit
- VE:6
- Bezeichnung:Hink's TNM-FH medium with L-glutamine lactalbumin hydrolysate, and yeastolate, without insect hemolymph
Spezifikationen
Über diesen Artikel
Hink's TNM-FH medium is a modified Grace's formulation containing lactalbumin hydrolysate (LAH) and ultrafiltered yeastolate, and was originally developed for the optimal propagation of cabbage looper, Tricoplusi in cells.