Stabiliser for calibrators and controls, SeramunStab® STD
Catalog # APOSBIL3120-100ML
Supplier: Apollo Scientific
CAS Number:
Specifications
- VE:100 mL
- Bezeichnung:SeramunStab® STD Stabiliser for Calibrators and controls
- Seq:01
Specifications
About this item
Liquid product yellow odourless pH-value 7,3 to 7,5 Boiling point 102 °C. Stabiliser for complex mixtures of biomolecules.