Capillaries for waters LC systems
HEWL8005-0812
: Agilent
- Ø int.:0,005" mm
- Bezeichnung:Capillary, red cap, PEEK
- Länge:0,52 m
- VE:1
These capillaries and tubing are made from ultra-clean stainless steel and are suitable for Waters HPLC systems. They are ideal for most standard applications.
- Reliable high-pressure sealing
- Smooth inner surface for the lowest backpressure
- Precision cutting for burr-free, square-cut ends without inner-diameter distortion
- Both ends accept standard fittings
- Predefined lengths for specific path locations to avoid peak broadening
- Individually tested to ensure reproducibility
Used with the right fittings, they are ideal for LC system plumbing, providing an inert surface, tight, leak-free connections, and zero dead volumes.