ContactUsLinkComponent
Alignment interfaces and cassettes
Catalog # HEWLG7100-60400
Supplier:  Agilent
Alignment interfaces and cassettes
Catalog # HEWLG7100-60400
Supplier:  Agilent
Specifications
  • Bezeichnung:
    Alignment interface for CE-MS capillary 360 µm Ext.Ø for G7100 capillary electrophoresis
  • VE:
    1
Specifications
About this item
These alignment interfaces are an integral part of the Agilent diode-array detection system.
  • The quick-change cassette allows capillary exchange in less than a minute
  • The cassette and interfaces accept all commercially available capillaries (~365 µm od)
These interfaces contain optical slits which are precisely matched to the capillary inner diameter for optimized sensitivity and linear detection range. In combination with the capillary cassette, alignment interfaces simplify capillary exchange, protect the fragile detection window and ensure exact alignment of the window in the detector.
Caution: The color code of the alignment interface must match with the color code of the capillary's built-in alignment stopper.