AuftragsverfolgungKontakt
Organic acids solution kit
 :   Agilent
Organic acids solution kit
 :  Agilent
Organic acids solution kit
The Organic Acids Solutions Kit contains capillaries, buffers, an organic acids standard and instructions complete with a sample electropherogram to help you begin organic acids analyses immediately.
  • Ideal for the analysis of short alkyl-chain carboxylic acids
  • Analyze food applications, including beer, wine and juice
  • Use for chemical applications, including raw materials and drug formulations
Employing an indirect UV detection agent optimized for organic acids, the methodology is simple, sensitive, and provides accurate quantitative analysis. Suited for the analysis of organic acids in a wide range of matrices, it is especially useful for determination of organic acids in beverages, and food.
Product Family Options
  • Bezeichnung
    VE
  • Organic acids solutions kit
    1 KIT
    1 * 1 KIT
    1 * 1 KIT - 913,00 €
    913,00 €
    1 * 1 KIT
       913,00 €
  • Buffer, organic acid, 250 ml, pH 5,6
    250 mL
    1 * 250 mL
    1 * 250 mL - 218,00 €
    218,00 €
    1 * 250 mL
       218,00 €
  • Organic acids test sample, 20 ml
    20 mL
    1 * 20 mL
    1 * 20 mL - 336,00 €
    336,00 €
    1 * 20 mL
       336,00 €