AuftragsverfolgungKontakt
Retroviral gene expression system, ViraPort | Avantor
Retroviral gene expression system, ViraPort
Artikelnr.
Retroviral gene expression system, ViraPort
Artikelnr.
Anbieter:  
CAS Nummer:  
Retroviral gene expression system, ViraPort
CAS Nummer:  
Product Details & Documents
The ViraPort retroviral gene expression system is a comprehensive line of gene delivery products providing transduction efficiencies approaching 100% in a wide range of mitotic cells.
  • Powerful system for functional cloning
  • Copy number controlled by multiplicity of infection
  • Create high-titer retroviral virions for library screening
Achtung! Bitte beachten: For Research Use Only. Not for Use in Diagnostic Procedures.