Connecting capillary, VWD
Capillary, S/E fittings, Stainless steel, 0,17 mm, 0,12 m
Artikelnr. HEWLG1314-87302
Anbieter: Agilent
Springe zu:
Product Details & Documents
A comprehensive range of capillaries and fittings are available to protect the integrity of the chromatographic results.
- Best fit to detector flow cell for optimum chromatographic performance
- Increased usability without scarifying durability
- Tight and leak free connection
Spezifikationen
- Ø int.:0,17 mm
- Bezeichnung:Capillary, S/E fittings, Stainless steel
- Länge:0,12 m