Überspringen zu Kopf-Ebene
Überspringen zu Hauptinhalt
Überspringen zu Fußzeile
Erweiterte Suche
Erweiterte Suche
Sicherheitsdatenblatt (SDS)
Zertifikate
Herstellerdaten
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Heim
Proteine und Peptide
Recombinant Epstein-Barr Virus (HHV-4) p18
Recombinant Epstein-Barr Virus (HHV-4) p18
Recombinant Epstein-Barr Virus (HHV-4) p18
Catalog # APOSBITP1916-1MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Recombinant Epstein-Barr Virus (HHV-4) p18
Catalog # APOSBITP1916-1MG
Supplier:
Apollo Scientific
CAS Number:
Specifications
VE:
1 mg
Protein/Peptid-Typ:
Rekombinant
Proteinsynonym:
EBV p18
Protein/Peptid-Name:
Epstein-Barr Virus (HHV-4) p18
Specifications
About this item
Product Details
Product Details