Recombinant Toxoplasma Gondii ROP4 RH2
Catalog # APOSBITP1853-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- VE:1 mg
- Protein/Peptid-Typ:Rekombinant
- Proteinsynonym:Toxoplasma ROP4 RH2
- Protein/Peptid-Name:Toxoplasma Gondii ROP4 RH2