Order Entry
Austria
ContactUsLinkComponent
 
Recombinant Hepatitis A Virus VP4-VP2
Catalog # APOSBITP1819-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Hepatitis A Virus VP4-VP2
Catalog # APOSBITP1819-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • VE:
    1 mg
  • Protein/Peptid-Typ:
    Rekombinant
  • Proteinsynonym:
    HAV VP4-VP2
  • Protein/Peptid-Name:
    Hepatitis A Virus VP4-VP2

Specifications

About this item