Recombinant Chlamydia Trachomatis HSP70 (462-503) (from CHO cells)
Supplier: Apollo Scientific
Certificates
About this item
Specifications
- Protein/Peptid-Typ:Rekombinant
- Ursprung:CHO cells
- Proteinsynonym:Chlamydia HSP70
- Protein/Peptid-Name:Chlamydia Trachomatis HSP70 (462-503)
- Sequence:462-503
Specifications
Frequently Bought Together