0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Proteine und Peptide
Recombinant Herpes Simplex Virus-8 Mosaic
Recombinant Herpes Simplex Virus-8 Mosaic
APOSBITP1740-1MG
:
Apollo Scientific
:
Recombinant Herpes Simplex Virus-8 Mosaic
APOSBITP1740-1MG
:
Apollo Scientific
:
VE:
1 mg
Protein/Peptid-Typ:
Rekombinant
Proteinsynonym:
HSV-8 Mosaic
Protein/Peptid-Name:
Herpes Simplex Virus-8 Mosaic