Recombinant SARS Associated Coronavirus Envelo
Catalog # APOSBITP1732-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- VE:1 mg
- Protein/Peptid-Typ:Rekombinant
- Proteinsynonym:SARS Envelope
- Protein/Peptid-Name:SARS Associated Coronavirus Envelo