Order Entry
Austria
ContactUsLinkComponent
2-Chlor-5-iodbenzotrifluorid 97%
2-Chlor-5-iodbenzotrifluorid 97%
Catalog # APOSPC520562-100G
Supplier:  Apollo Scientific
CAS Number:  
2-Chlor-5-iodbenzotrifluorid 97%
Catalog # APOSPC520562-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Packung:
    Bottle
  • VE:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :306.4514 g/mol
Storage Temperature:Raumtemperatur
CAS Number:260355-20-2
Formula:C₇H₃ClF₃I
MDL Number:MFCD09037801