Express Eingabe
Austria
AuftragsverfolgungKontakt
Adipohydrazid 97%

Adipohydrazid 97%

New Product
Supplier:   Apollo Scientific
Adipohydrazid 97%
Supplier:  Apollo Scientific
New Product
Certificates
Adipohydrazid 97%

3 Options Available Below

Product Details & Documents
Synonyms:  Adipyl hydrazide
  • CAS Number:1071-93-8
  • Formula:C₆H₁₄N₄O₂
  • MDL Number:MFCD00007614
  • Molecular Weight:174.2 g/mol
  • Melting Pt:180…182 °C
  • Storage Temperature:Raumtemperatur
Product Family Options

Product Information

  • Packung
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 21,80 €
    21,80 €
    1 * 25 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 24,60 €
    24,60 €
    1 * 100 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 254,00 €
    254,00 €
    1 * 2,5 kg
       254,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.