3-Methyl-1-phenyl-2-pyrazolin-5-on
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:89-25-8
:
Formula:C₁₀H₁₀N₂O
MDL Number:MFCD00003138
:174.2 g/mol
Boiling Pt:287 °C/265mmHg
Melting Pt:126…128 °C
Density:1,12 g/cm³ (25 °C)
Storage Temperature:Raumtemperatur
EINECS:201-891-0
:GHS07
Frequently Bought Together