AuftragsverfolgungKontakt
AMPSO ≥98%
AMPSO ≥98%
AMPSO ≥98%
Biological buffer - useful range 8.3-9.7.
 :  N-(1,1-Dimethyl-2-hydroxyethyl)-3-amino-2-hydroxypropanesulphonic acid
  •  :68399-79-1
  •  :C₇H₁₇NO₅S
  •  :MFCD00041777
  •  :227.28 g/mol
  •  :Raumtemperatur
Product Family Options
  • Packung
    VE
  • Bottle
    1 * 25 g
    1 * 25 g - 102,00 €
    102,00 €
    1 * 25 g
       102,00 €
    New Product
    New Product
  • Bottle
    100 g
    1 * 100 g
    1 * 100 g - 262,00 €
    262,00 €
    1 * 100 g
       262,00 €