Desoxy-Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]desoxycholamid)
Artikelnr. APOSBIB1156-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Packung:Bottle
- VE:250 mg
- Seq:2590
Specifications
About this item
:
:862.05 g/mol
:GHS07
Storage Temperature:Kühlschrank
CAS Number:86303-23-3
Formula:C₄₂H₇₅N₃O₁₅
MDL Number:MFCD00161482