Human recombinant glypican 3 (from HEK293)
- Konjugation:Unconjugated
- Protein/Peptid-Typ:Rekombinant
- Ursprung:HEK293 cells
- Spezies:Human
- Lagerbedingungen:–20 °C
- Endotoxingehalt:Less than 0,1 EU per μg by the LAL method.
- Biologische Aktivität:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human FGF basic, Tag Free (ACRBGP3-H5258-100) at 2 μg/ml (100 μl/well)can bind Human Glypican 3, Fc Tag (ACRBGP3-H5258-100) with a linear range of 4 - 63 ng/ml (QC tested). Measured by its binding ability in a functional ELISA. Immobilised Human Glypican 3, Fc Tag, low endotoxin (ACRBGP3-H5258-100) at 0,5 μg/ml (100 μl/well) can bind Anti-Human GPC3 Mab with a linear range of 0,8 - 6 ng/ml (Routinely tested). 293 cells were transfected with anti-GPC3-scFv and RFP tag. 2e5 of the cells were first stained with B. Human Glypican 3, Fc Tag, low endotoxin (ACRBGP3-H5258-100, 3 μg/ml) and C. human Fc tag protein control, followed by FITC-conjugated anti-human IgG Fc antibody. A. Non-transfected 293 cells and C. human Fc tag protein control were used as negative control. RFP was used to evaluate CAR (anti-GPC3-scFv) expression and FITC was used to evaluate the binding activity of Human Glypican 3, Fc Tag, low endotoxin (ACRBGP3-H5258-100).
- Gen-ID:P51654-1
- Frei von Trägern:Y
- Frei von Proteasen:Y
- Tierfrei:Y
- Proteinsynonym:GPC3|SGBS|SGB|Glypican-3|SDYS |MXR7|SGBS1|OCI5|GTR2-2|DGSX
- Protein/Peptid-Name:Glypican 3
- Purity:>90%
- Molecular weight:38,1 kDa and 49,3 kDa
- Endotoxinspiegel:Low
- Zusammensetzung:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Frei von Nukleasen:Y
- Transporttemperatur:Powder, RT
Frequently Bought Together