Anti-S1 protein NTD HEK293 monoclonal antibody
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Type: Primary
Antigen: S1 protein NTD
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: HEK293
Isotype: Human IgG1/kappa
Reactivity:
Specifications
- Catalog No:
- ACRBSPD-M121-1MG
- ACRBSPD-M121-100UG
- Antigen-Symbol:
- S1 protein NTD
- S1 protein NTD
- Antigen-Name:
- S1 protein NTD
- S1 protein NTD
- Antigen-Synonyme:
- S1 protein NTD,Spike protein S1 NTD,BetaCoV S1-NTD,COVID-19
- S1 protein NTD,Spike protein S1 NTD,BetaCoV S1-NTD,COVID-19
- Antikörper-Typ:
- Primär
- Primär
- Klonalität:
- Monoklonal
- Monoklonal
- Konjugation:
- Unconjugated
- Unconjugated
- Wirt:
- HEK293
- HEK293
- ELISA:
- Yes
- Yes
- Form:
- Powder
- Powder
- Reinigung:
- Affinity purified
- Affinity purified
- Kreuzadsorption:
- The cross-reactivity with other coronaviruses has not been tested yet.
- The cross-reactivity with other coronaviruses has not been tested yet.
- Lagertemperatur:
- For long term storage, the product should be stored at lyophilised state at −20°C or lower.
- For long term storage, the product should be stored at lyophilised state at −20°C or lower.
- Transporttemperatur:
- RT
- RT