Human recombinant TROP-2 (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Human TROP-2 / TACSTD2 Protein, Fc Tag, ACROBiosystems
Specifications
- Konjugation:Unconjugated
- Protein/Peptid-Typ:Rekombinant
- Ursprung:HEK293 cells
- Spezies:Human
- Tag sequence:Fc Tag
- Lagerbedingungen:–20 °C
- Endotoxingehalt:Less than 1,0 EU per μg by the LAL method
- Gen-ID:P09758-1
- Proteinsynonym:M1S1|GA733-1|TACSTD2|TROP2
- Protein/Peptid-Name:TROP-2
- Purity:>95% as determined by SDS-PAGE
- Molecular weight:54,3 kDa
- Endotoxinspiegel:Low
- Zusammensetzung:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
- Transporttemperatur:RT