M’Fadyean Stain solution, Sigma-Aldrich®
Artikelnr.
Product Details & Documents
M′Fadyean Stain is used for capsule staining of Bacillus anthracis.
Synonyme: Polychrom Methylene Blue Solution according to M'Fadyean
- Lagertemperatur:Raumtemperatur



