Überspringen zu Kopf-Ebene
Überspringen zu Hauptinhalt
Überspringen zu Fußzeile
Erweiterte Suche
Erweiterte Suche
Sicherheitsdatenblatt (SDS)
Zertifikate
Herstellerdaten
0
0,00 €
Order Entry
Login
/
Registrieren
Products
Services
Digital Solutions
Applications
Support
About
Express Eingabe
Bestellstatus
Ihr Konto
Warenkorb
Austria
Express Eingabe
Bestellstatus
Ihr Konto
Warenkorb
ContactUsLinkComponent
Heim
Gram-Nicolle Kit
Gram-Nicolle Kit To differentiate Gram-positive from Gram-negative Bacteria
Gram-Nicolle Kit To differentiate Gram-positive from Gram-negative Bacteria
Catalog # RALA364320-0000
Supplier:
RAL
CAS Number:
Certificates
Expand image
Gram-Nicolle Kit To differentiate Gram-positive from Gram-negative Bacteria
Catalog # RALA364320-0000
Supplier:
RAL
CAS Number:
Certificates
Specifications
VE:
1 KIT
Seq:
0075
Specifications
About this item
Product Details
Documentation
:
:
1
Product Details
Documentation