AuftragsverfolgungKontakt
Universal sorbent rolls | Avantor
Universal sorbent rolls
Universal sorbent rolls, heavyweight, perforated and bonded, 192 L, 44000×4800 mm
Artikelnr. BRDY813828
Anbieter:   Brady
Universal sorbent rolls
Artikelnr. BRDY813828
Anbieter:  Brady
Universal sorbent rolls
Universal sorbent rolls
Universal sorbent rolls, heavyweight, perforated and bonded, 192 L, 44000×4800 mm
Preis
213,00 €
Einloggen für Verfügbarkeit
1 * 2 Rolle
Product Details & Documents
These grey coloured medium weight, bonded and perforated universal sorbent rolls absorbs industrial liquids including oils, water, solvents, coolants.
  • Made of one-ply, meltblown, bonded
  • Strong and durable with a superior finish with less lint for improved abrasion resistance
Ideal to cover a large surface area for fast clean up of spills, leaks and drips. Suited for industrial applications where low cost and high absorbency are required.
Spezifikationen
  • Bezeichnung:
    Universal sorbent rolls, heavyweight, perforated and bonded
  • L×B:
    44000×4800 mm
  • Absorptionsvermögen:
    192 L