Universal sorbent pads
Universal sorbent pads, heavyweight, perforated and bonded, 106 L, 400×500 mm
Artikelnr. BRDY813809
Anbieter: Brady
Springe zu:
Product Details & Documents
These grey coloured heavy weight, bonded and perforated universal sorbent pad absorbs industrial liquids including oils, water, solvents, coolants.
- One sided spunbound for extra strength and less lint
- Made of two-ply, spunbound/meltblown, bonded
Perforated to help minimise waste and lower disposal costs. Ideal sorbent for dealing with leaks and drips in the workplace and also suited for general industrial maintenance and spill clean up.
Spezifikationen
- Bezeichnung:Universal sorbent pads, heavyweight, perforated and bonded
- L×B:400×500 mm
- Absorptionsvermögen:106 L