Human Recombinant PDGF-BB (Animal free) (from E. coli)
Lieferant: Shenandoah Biotechnology
Zertifikate
Über diesen Artikel
Receptor activator of nuclear factor kappa-B Ligand (RANK Ligand) is a cell-bound marker related to the tumor necrosis factor (TNF) family of proteins. RANK Ligand plays a critical role in bone metabolism and osteoclast differentiation. T cell expression of RANK Ligand promotes dendritic cell maturation.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Delivery: Certificate of Analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Caution: For research use only.
Spezifikationen
- Konjugation:Unconjugated
- Protein-Funktion:Cytokine
- Protein/Peptid-Typ:Rekombinant
- Ursprung:E. coli
- Spezies:Human
- Lagerbedingungen:−20 °C
- Endotoxingehalt:Endotoxin LAL (EU/µg) ≤0.1
- Biologische Aktivität:Bioactivity assay: NR6R-3T3 cell proliferation, ED50 ≤20 ng/ml, bioactivity ≥5,0×10⁴ units/mg
- Gen-ID:P01127
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Frei von Endotoxinen:N
- Frei von Trägern:Y
- Frei von Proteasen:N
- Tierfrei:Y
- Proteinsynonym:ODGF|GDGF
- UniProtKB:P01127
- Protein/Peptid-Name:PDGF-BB
- Purity:≥95%
- Molecular weight:12,4/24,9 kDa
- Sequence:MSLGSLTIAE PAMIAECKTR TEVFEISRRL IDRTNANFLV WPPCVEVQRC SGCCNNRNVQ CRPTQVQLRP VQVRKIEIVR KKPIFKKATV TLEDHLACKC ETVAAARPVT
- Endotoxinspiegel:Low
- Zusammensetzung:10 mM sodium phosphate, pH 7,5
- Frei von Nukleasen:N
- Sorte:RUO
- Transporttemperatur:RT
- Validierte Anwendungen:Bioassay
Spezifikationen
Zugehörige Informationen
Literature reference - PubMed ID 25121729 Literature reference - PubMed ID 25287123
Weitere Produktdetails
Frequently Bought Together