
Dabsylchlorid 98,0-102,0% (durch argentometrische Titration), LiChropur™ Derivatisierungsreagenz, für die HPLC, Supelco®
89802-500MG
: Merck
:
- VE:500 mg
- Seq:2725
- Appearance (Color):Red to Very Dark Red and Purple to Very Dark Purple and Red-Purple
- Appearance (Form):Powder to Crystals
- Argentometr. Titration:98.0 - 102.0 %
- Infrared Spectrum:Conforms to Structure
:
:323.80 g/mol
:185 °C (Zers., Lit.)
:260-235-1
:3261
:8,II
:GHS05
:56512-49-3
:C₁₄H₁₄ClN₃O₂S
:MFCD00007444