Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organische Standards
Thiochrome, analytical standard, Supelco®
Thiochrome, analytical standard, Supelco®
Thiochrome, analytical standard, Supelco®
Catalog # T7891-100MG
Supplier:
Merck
CAS Number:
Expand image
Thiochrome, analytical standard, Supelco®
Catalog # T7891-100MG
Supplier:
Merck
CAS Number:
Specifications
VE:
100 mg
Normen:
analytical standard
CAS Nr.:
92-35-3
Bezeichnung:
Thiochrome
Bezeichnung:
Thiochrome
Specifications
About this item
Product Details
Product Details