0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Standards
Organische Standards
Tetrachlorobisphenol A, analytical standard, Supelco®
Tetrachlorobisphenol A, analytical standard, Supelco®
73081-10MG
:
Merck
:
Tetrachlorobisphenol A, analytical standard, Supelco®
73081-10MG
:
Merck
:
VE:
10 mg
Normen:
analytical standard
CAS Nr.:
79-95-8
Bezeichnung:
Tetrachlorobisphenol A
Bezeichnung:
Tetrachlorobisphenol A