0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Standards
Organische Standards
AMOZ, analytical standard, VETRANAL™, Supelco®
AMOZ, analytical standard, VETRANAL™, Supelco®
Catalog # 33349-50MG-R
Supplier:
Merck
CAS Number:
Expand image
AMOZ, analytical standard, VETRANAL™, Supelco®
Catalog # 33349-50MG-R
Supplier:
Merck
CAS Number:
Specifications
VE:
50 mg
Normen:
analytical standard
CAS Nr.:
43056-63-9
Bezeichnung:
AMOZ
M-Clarity Qualitätsstufe:
MQ100
Bezeichnung:
AMOZ
Specifications
About this item
Product Details
Product Details