IQ/OQ certificaton for gas generators
Catalog # PEAK09-2100
Supplier: PEAK SCIENTIFIC
Specifications
- Bezeichnung:Installation and Operational Qualification (IQ/OQ)
- VE:1
Specifications
About this item
With customer service and support in mind a comprehensive Installation and Operational Qualification certification has been developed, for customers who require this additional certified assurance for their analytical applications.
- IQ/OQ is tailored to the specific generator
- Generator compliance certification for regulated applications
- Carried out at initial installation or equipment relocation
IQ is the pre-installation detail of the equipment, its component parts and its location onsite.
OQ is to verify that the equipment functions according to the generator’s specifications.
This provides a cost-effective and efficient solution to achieve and maintain compliance. In addition to our highly trained Service Engineer completing the Installation and Operational validation, we will also provide full certification to document this process, ensuring accurate proof of compliance.