Supelco® Organic Reference Standard, α-Arbutin
03338-100MG
: Merck
:
- VE:100 mg
- Normen:Analytical standard
- CAS Nr.:84380-01-8
- Bezeichnung:α-Arbutin
- Lagerbedingungen:2...8 °C
- Shipping conditions:STD
- MDL No.:MFCD09838262
- M-Clarity Qualitätsstufe:MQ100
- Synonym(s):4-Hydroxyphenyl α-ᴅ-glucopyranoside; Hydroquinone O-α-ᴅ-glucopyranoside
- Formel:C₁₂H₁₆O₇
- Molecular weight:272,25
- Lagertemperatur:2...8 °C
- Flammpunkt:Not applicable