Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, 2,6-Dimethoxyphenol, analytical standard, Supelco®
Catalog # 53877-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 2,6-Dimethoxyphenol, analytical standard, Supelco®
Catalog # 53877-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • VE:
    100 mg
  • Normen:
    Analytical standard
  • CAS Nr.:
    91-10-1
  • Bezeichnung:
    2,6-Dimethoxyphenol, ≥98,0% (GC)
  • Melting point:
    50...57 °C (lit.)
    53...57 °C
  • Formel:
    C₈H₁₀O₃
  • Bezeichnung:
    2,6-Dimethoxyphenol
  • Siedepunkt:
    261 °C (lit.)
  • Form:
    Neat

Specifications

About this item

2,6-Dimethoxyphenol can find applications mainly as a substrate of laccases catalysis and also determine its activity in aqueous solution. It can also be used as a potential marker compound for emission of smoke in the atmosphere from wood.

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.