ContactUsLinkComponent
n-Paraffin Mix C18, C20, C22, C24, certified reference material, Supelco®
n-Paraffin Mix C18, C20, C22, C24, certified reference material, Supelco®
Catalog # 47108.
Supplier:  Merck
CAS Number:  
n-Paraffin Mix C18, C20, C22, C24, certified reference material, Supelco®
Catalog # 47108.
Supplier:  Merck
CAS Number:  
Specifications
  • Konzentration:
    2 % (w/w) each component in octane
  • VE:
    1
  • Bezeichnung:
    n-Paraffin Mix C18, C20, C22, C24