Trace Metals 2 in Seawater - Whole Volume - WP, Proficiency Testing Material, Supelco®
Catalog # PE3189-500ML
Supplier: Merck
CAS Number:
Specifications
- VE:500 mL
- Bezeichnung:Trace Metals 2 in Seawater - Whole Volume - WP
- Typ:Proficiency Testing Material