Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Umweltanalyse-Standards
Bromate and bromide - WS, proficiency testing material, Supelco®
Bromate and bromide - WS, proficiency testing material, Supelco®
Bromate and bromide - WS, proficiency testing material, Supelco®
Catalog # PE1361-2ML
Supplier:
Merck
CAS Number:
Expand image
Bromate and bromide - WS, proficiency testing material, Supelco®
Catalog # PE1361-2ML
Supplier:
Merck
CAS Number:
Specifications
VE:
2 mL
Bezeichnung:
Bromate and Bromide - WS
Typ:
Proficiency Testing Material
Specifications
About this item
Product Details
Product Details