Supelco® Organic Reference Standard, 2-Stearoylglycerol
36484-25MG
: Merck
:
- VE:25 mg
- Normen:Analytical standard
- CAS Nr.:621-61-4
- Bezeichnung:2-Stearoylglycerol
- Lagerbedingungen:–70 °C
- Shipping conditions:DICE
- MDL No.:MFCD25974054
- M-Clarity Qualitätsstufe:MQ100
- Synonym(s):β-Monostearin; 2-Monostearin; 2-Monostearoylglycerol; 2-Octadecanoylglycerol; Glycerol β-monostearate; Octadecanoic acid 2-hydroxy-1-(hydroxymethyl)ethyl ester
- Formel:C₂₁H₄₂O₄
- Molecular weight:358,56
- Lagertemperatur:−70 °C
- Flammpunkt:Not applicable