ContactUsLinkComponent
Cell-based assay calcein AM
  CAYM400146-1
 :  
Cell-based assay calcein AM
  CAYM400146-1
 :  
  • VE:
    1
  • Bezeichnung:
    Cell-based assay calcein AM
 : This vial contains 50 µl of 1 mM calcein AM solution in DMSO.