Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Umweltanalyse-Standards
BNAs in Soil, (CRM), Supelco®
BNAs in Soil, (CRM), Supelco®
BNAs in Soil, (CRM), Supelco®
Catalog # SQC003-40G
Supplier:
Merck
CAS Number:
Expand image
BNAs in Soil, (CRM), Supelco®
Catalog # SQC003-40G
Supplier:
Merck
CAS Number:
Specifications
VE:
40 g
Bezeichnung:
BNAs in Soil
Typ:
Certified reference material
Specifications
About this item
Product Details
Product Details