Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Registrieren
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Umweltanalyse-Standards
Acidity (as CaCO₃), concentrate, (CRM), Supelco®
Acidity (as CaCO₃), concentrate, (CRM), Supelco®
Acidity (as CaCO₃), concentrate, (CRM), Supelco®
Catalog # QC1319-20ML
Supplier:
Merck
CAS Number:
Expand image
Acidity (as CaCO₃), concentrate, (CRM), Supelco®
Catalog # QC1319-20ML
Supplier:
Merck
CAS Number:
Specifications
VE:
20 mL
Bezeichnung:
Acidity (as CaCO3), concentrate
Typ:
Certified reference material
Specifications
About this item
Product Details
Product Details